tert-Butyl [2-(methylamino)ethyl]carbamate hydrochloride 95%

Supplier: Apollo Scientific
Warning

OR0802-1G OR0802-5G OR0802-250MG
APOSOR0802-1GEA 45.1 EUR
APOSOR0802-1G APOSOR0802-5G APOSOR0802-250MG
tert-Butyl [2-(methylamino)ethyl]carbamate hydrochloride 95%
tert-Butyl [2-(methylamino)ethyl]carbamate hydrochloride
Formula: C₈H₁₈N₂O₂·ClH
Melting Pt: 129…130 °C
Storage Temperature: Ambient
MDL Number: MFCD06796281
CAS Number: 202207-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR