4-Amino-2-chloropyridine

Supplier: Apollo Scientific
Warning

Synonyms: 2-Chloro-4-aminopyridine

OR10317-100G OR10317-500G
APOSOR10317-100GEA 60.9 EUR
APOSOR10317-100G APOSOR10317-500G
4-Amino-2-chloropyridine
4-Amino-2-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 300
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00060089
CAS Number: 14432-12-3
EINECS: 238-403-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR