4-(Cyclohexylcarbamoyl)phenylboronic acid

Supplier: Apollo Scientific
Warning

OR10430-1G OR10430-5G OR10430-25G
APOSOR10430-1GEA 52.2 EUR
APOSOR10430-1G APOSOR10430-5G APOSOR10430-25G
4-(Cyclohexylcarbamoyl)phenylboronic acid
4-(Cyclohexylcarbamoyl)phenylboronic acid
Formula: C₁₃H₁₈BNO₃
Melting Pt: 174…178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411956
CAS Number: 762262-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR