1,4-Phenylenediphosphonic acid

Supplier: Apollo Scientific
Danger

OR10672-1G OR10672-250MG
APOSOR10672-1GEA 116 EUR
APOSOR10672-1G APOSOR10672-250MG
1,4-Phenylenediphosphonic acid
1,4-Phenylenediphosphonic acid
Formula: C₆H₈O₆P₂
Melting Pt: >260 °C
Storage Temperature: Ambient
MDL Number: MFCD00475582
CAS Number: 880-68-2
UN: 3261
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR