Diethyl (2-bromoethyl)phosphonate 97%

Supplier: Apollo Scientific
Warning

OR10692-100G OR10692-25G
APOSOR10692-100GEA 422 EUR
APOSOR10692-100G APOSOR10692-25G
Diethyl (2-bromoethyl)phosphonate 97%
Diethyl (2-bromoethyl)phosphonate
Formula: C₆H₁₄BrO₃P
MW: 245.06 g/mol
Boiling Pt: 98…100 °C/1mmHg
Density: 1.348 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000239
CAS Number: 5324-30-1
EINECS: 226-193-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR