2-Aminobenzyl alcohol

Supplier: Apollo Scientific
Warning

OR10943-250G OR10943-50G
APOSOR10943-250GEA 116 EUR
APOSOR10943-250G APOSOR10943-50G
2-Aminobenzyl alcohol
2-Aminobenzyl alcohol
Formula: C₇H₉NO
Boiling Pt: 160 °C/10mmHg
Melting Pt: 81…84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007749
CAS Number: 5344-90-1
EINECS: 226-293-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR