N-(2,4-Dichloro-6-methylphenyl)-N'-methylthiourea

Supplier: Apollo Scientific

OR110053-1G
APOSOR110053-1GEA 186 EUR
APOSOR110053-1G
N-(2,4-Dichloro-6-methylphenyl)-N'-methylthiourea
N-(2,4-Dichloro-6-methylphenyl)-N'-methylthiourea
Formula: C₉H₁₀Cl₂N₂S
MW: 249,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24850054
CAS Number: 1440535-56-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR