7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid

Supplier: Apollo Scientific
Warning

OR110102-1G
APOSOR110102-1GEA 0 EUR
APOSOR110102-1G
7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid
7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid
Formula: C₈H₅BrN₂O₂
Melting Pt: 234…236 °C
Storage Temperature: Ambient
MDL Number: MFCD22989340
CAS Number: 1019018-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR