(4-Amino-3-bromophenyl)acetonitrile

Supplier: Apollo Scientific

OR110126-1G OR110126-250MG OR110126-500MG OR110126-2.5G OR110126-5G
APOSOR110126-1GEA 102 EUR
APOSOR110126-1G APOSOR110126-250MG APOSOR110126-500MG APOSOR110126-2.5G APOSOR110126-5G
(4-Amino-3-bromophenyl)acetonitrile
(4-Amino-3-bromophenyl)acetonitrile
Formula: C₈H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD08275180
CAS Number: 882855-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR