3-Bromoimidazo[1,2-a]pyridine-7-carbohydrazide

Supplier: Apollo Scientific
Warning

OR110147-1G OR110147-250MG
APOSOR110147-1GEA 743 EUR
APOSOR110147-1G APOSOR110147-250MG
3-Bromoimidazo[1,2-a]pyridine-7-carbohydrazide
3-Bromoimidazo[1,2-a]pyridine-7-carbohydrazide
Formula: C₈H₇BrN₄O
Storage Temperature: Ambient
MDL Number: MFCD21602378
CAS Number: 1427460-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR