Ethyl-6-methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Supplier: Apollo Scientific
Warning

OR11018-1G OR11018-5G
APOSOR11018-1GEA 56.7 EUR
APOSOR11018-1G APOSOR11018-5G
Ethyl-6-methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate
Ethyl-6-methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate
Formula: C₁₄H₁₆N₂O₃
Melting Pt: 208 °C
Storage Temperature: Ambient
MDL Number: MFCD00124778
CAS Number: 5395-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR