Ethyl-3-amino-1-benzothiophene-2-carboxylate

Supplier: Apollo Scientific

OR110192-5G OR110192-25G
APOSOR110192-5GEA 88.6 EUR
APOSOR110192-5G APOSOR110192-25G
Ethyl-3-amino-1-benzothiophene-2-carboxylate
Ethyl-3-amino-1-benzothiophene-2-carboxylate
Formula: C₁₁H₁₁NO₂S
Melting Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00661903
CAS Number: 34761-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR