Pyrazine-2-carboxylic acid

Supplier: Apollo Scientific
Warning

OR11036-500G
APOSOR11036-500GEA 72.5 EUR
APOSOR11036-500G
Pyrazine-2-carboxylic acid
Pyrazine-2-carboxylic acid
Formula: C₅H₄N₂O₂
Melting Pt: 222…225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006130
CAS Number: 98-97-5
EINECS: 202-718-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR