6-Bromo-3-nitroimidazo[1,2-a]pyridine

Supplier: Apollo Scientific

OR110403-1G
APOSOR110403-1GEA 97.3 EUR
APOSOR110403-1G
6-Bromo-3-nitroimidazo[1,2-a]pyridine
6-Bromo-3-nitroimidazo[1,2-a]pyridine
Formula: C₇H₄BrN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09864963
CAS Number: 64064-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR