Methyl-4-(hydroxymethyl)benzoate

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 4-(hydroxymethyl)benzoate

OR110419-10G OR110419-5G OR110419-25G OR110419-100G
APOSOR110419-10GEA 24.8 EUR
APOSOR110419-10G APOSOR110419-5G APOSOR110419-25G APOSOR110419-100G
Methyl-4-(hydroxymethyl)benzoate
Methyl 4-(hydroxymethyl)benzoate
Formula: C₉H₁₀O₃
Melting Pt: 46…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00009984
CAS Number: 6908-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR