3-[(5-Bromopyridin-2-yl)oxy]benzaldehyde

Supplier: Apollo Scientific

OR110555-1G OR110555-5G
APOSOR110555-1GEA 371 EUR
APOSOR110555-1G APOSOR110555-5G
3-[(5-Bromopyridin-2-yl)oxy]benzaldehyde
3-[(5-Bromopyridin-2-yl)oxy]benzaldehyde
Formula: C₁₂H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD17169531
CAS Number: 1427460-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR