2-Methyl-1,3-benzoxazol-6-amine

Supplier: Apollo Scientific
Danger

OR110719-1G OR110719-5G
APOSOR110719-1GEA 55.1 EUR
APOSOR110719-1G APOSOR110719-5G
2-Methyl-1,3-benzoxazol-6-amine
2-Methyl-1,3-benzoxazol-6-amine
Formula: C₁₁H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD03791181
CAS Number: 5676-60-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR