3-Bromoimidazo[1,2-a]pyridin-8-amine

Supplier: Apollo Scientific
Warning

OR110820-250MG
APOSOR110820-250MGEA 186 EUR
APOSOR110820-250MG
3-Bromoimidazo[1,2-a]pyridin-8-amine
3-Bromoimidazo[1,2-a]pyridin-8-amine
Formula: C₇H₆BrN₃
Storage Temperature: Ambient
MDL Number: MFCD17012030
CAS Number: 1232431-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR