5-(4-tert-Butylphenyl)-1-phenyl-1,2-dihydro-3H-1,2,4-triazole-3-thione

Supplier: Apollo Scientific

OR110850-1G
APOSOR110850-1GEA 371 EUR
APOSOR110850-1G
5-(4-tert-Butylphenyl)-1-phenyl-1,2-dihydro-3H-1,2,4-triazole-3-thione
5-(4-tert-Butylphenyl)-1-phenyl-1,2-dihydro-3H-1,2,4-triazole-3-thione
Formula: C₁₈H₁₉N₃S
Storage Temperature: Ambient
MDL Number: MFCD26936321
CAS Number: 1858255-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR