3-(4-Methylpiperazin-1-ylmethyl)benzylamine

Supplier: Apollo Scientific
Warning

Synonyms: 1-{3-[(4-Methylpiperazin-1-yl)methyl]phenyl}methanamine

OR111035-1G
APOSOR111035-1GEA 0 EUR
APOSOR111035-1G
3-(4-Methylpiperazin-1-ylmethyl)benzylamine
3-(4-Methylpiperazin-1-ylmethyl)benzylamine
Formula: C₁₃H₂₁N₃
Storage Temperature: Refrigerator
CAS Number: 515162-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR