(6-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid

Supplier: Apollo Scientific
Warning

OR111221-1G OR111221-5G OR111221-10G
APOSOR111221-1GEA 37.7 EUR
APOSOR111221-1G APOSOR111221-5G APOSOR111221-10G
(6-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid
(6-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid
Formula: C₉H₇BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06739269
CAS Number: 59128-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR