4-Nitro-1-indanone

Supplier: Apollo Scientific
Warning

OR111546-1G
APOSOR111546-1GEA 140 EUR
APOSOR111546-1G
4-Nitro-1-indanone
4-Nitro-1-indanone
Formula: C₉H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD06656901
CAS Number: 24623-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR