4-Chloro-1-naphthol

Supplier: Apollo Scientific
Warning

Synonyms: 4-Chloronaphthalen-1-ol, 1-Chloro-4-hydroxynaphthalene, 4-CN, Chloronaphthol

OR11309-25G OR11309-10G OR11309-100G
APOSOR11309-25GEA 132 EUR
APOSOR11309-25G APOSOR11309-10G APOSOR11309-100G
4-Chloro-1-naphthol
4-Chloro-1-naphthol
Formula: ClC₁₀H₆OH
Melting Pt: 120…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00003974
CAS Number: 604-44-4
EINECS: 210-068-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR