1-Bromo-2-(methoxymethyl)benzene 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Bromobenzyl methyl ether

OR11434-1G OR11434-5G OR11434-25G
APOSOR11434-1GEA 21.9 EUR
APOSOR11434-1G APOSOR11434-5G APOSOR11434-25G
1-Bromo-2-(methoxymethyl)benzene 97%
1-Bromo-2-(methoxymethyl)benzene
Formula: C₈H₉BrO
Boiling Pt: 75…80 °C/0,5mmHg
Storage Temperature: Ambient
MDL Number: MFCD03790887
CAS Number: 52711-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR