5-(Ethoxycarbonyl)-2-methoxybenzeneboronic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 3-borono-4-methoxybenzoate, 5-(Ethoxycarbonyl)-2-methoxyphenylboronic acid, 2-Borono-4-(ethoxycarbonyl)anisole

OR11693-1G OR11693-250MG OR11693-5G
APOSOR11693-1GEA 177 EUR
APOSOR11693-1G APOSOR11693-250MG APOSOR11693-5G
5-(Ethoxycarbonyl)-2-methoxybenzeneboronic acid 95%
5-(Ethoxycarbonyl)-2-methoxybenzeneboronic acid
Formula: C₁₀H₁₃BO₅
MW: 224.018 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800892
CAS Number: 957062-53-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR