3,5-Dimethoxyphenol

Supplier: Apollo Scientific
Warning

OR1170-1G OR1170-5G
APOSOR1170-1GEA 21.9 EUR
APOSOR1170-1G APOSOR1170-5G
3,5-Dimethoxyphenol
3,5-Dimethoxyphenol
Formula: C₈H₁₀O₃
Boiling Pt: 172 - 175/17 mm °C
Melting Pt: 40 - 45 °C
Flash Pt: 78 °C
MDL Number: MFCD00008388
CAS Number: 500-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR