6-(Pyrrolidin-1-yl)pyridine-2-carboxylic acid 97%

Supplier: Apollo Scientific
Warning

OR12215-1G
APOSOR12215-1GEA 0 EUR
APOSOR12215-1G
6-(Pyrrolidin-1-yl)pyridine-2-carboxylic acid 97%
6-(Pyrrolidin-1-yl)pyridine-2-carboxylic acid
Formula: C₁₀H₁₂N₂O₂
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD09064946
CAS Number: 450368-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR