4-Phenoxypiperidine hydrochloride

Supplier: Apollo Scientific
Warning

OR12597-1G OR12597-250MG
APOSOR12597-1GEA 29 EUR
APOSOR12597-1G APOSOR12597-250MG
4-Phenoxypiperidine hydrochloride
4-Phenoxypiperidine hydrochloride
Formula: C₁₁H₁₆ClNO
MW: 213.7038 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684106
CAS Number: 3413-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR