4-Hydroxy-3-nitroanisole

Supplier: Apollo Scientific
Warning

OR12652-100G OR12652-25G OR12652-5G
APOSOR12652-100GEA 143 EUR
APOSOR12652-100G APOSOR12652-25G APOSOR12652-5G
4-Hydroxy-3-nitroanisole
4-Hydroxy-3-nitroanisole
Formula: C₇H₇NO₄
Melting Pt: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00024247
CAS Number: 1568-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR