3-[(Pyrrolidin-3-yl)methyl]pyridine oxalate

Supplier: Apollo Scientific
Danger

OR12929-50MG
APOSOR12929-50MGEA 36.4 EUR
APOSOR12929-50MG
3-[(Pyrrolidin-3-yl)methyl]pyridine oxalate
3-[(Pyrrolidin-3-yl)methyl]pyridine oxalate
Formula: C₁₀H₁₄N₂·C₂H₂O₄
Storage Temperature: Ambient
MDL Number: MFCD04117738
CAS Number: 1018827-46-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR