Chromone-3-carboxaldehyde

Supplier: Apollo Scientific

Synonyms: 4-Oxo-4H-chromene-3-carbaldehyde

OR1305-100G
APOSOR1305-100GEA 179 EUR
APOSOR1305-100G
Chromone-3-carboxaldehyde
Chromone-3-carboxaldehyde
Formula: C₁₀H₆O₃
MW: 174.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014667
CAS Number: 17422-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR