4-Aminophenylboronic acid pinacol ester

Supplier: Apollo Scientific
Warning

OR13050-1G OR13050-25G OR13050-5G
APOSOR13050-1GEA 21.9 EUR
APOSOR13050-1G APOSOR13050-25G APOSOR13050-5G
4-Aminophenylboronic acid pinacol ester
4-Aminophenylboronic acid pinacol ester
Formula: C₁₂H₁₈BNO₂
Melting Pt: 165…171 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093721
CAS Number: 214360-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR