(±)-4-Chloromandelic acid

Supplier: Apollo Scientific
Warning

OR13218-100G OR13218-25G OR13218-5G
APOSOR13218-100GEA 50.9 EUR
APOSOR13218-100G APOSOR13218-25G APOSOR13218-5G
(±)-4-Chloromandelic acid
(±)-4-Chloromandelic acid
Formula: C₈H₇ClO₃
Melting Pt: 117…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00042724
CAS Number: 492-86-4
EINECS: 207-764-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR