tert-Butyl-4-formylpiperidine-1-carboxylate 97%

Supplier: Apollo Scientific
Warning

OR13630-10G
APOSOR13630-10GEA 21.9 EUR
APOSOR13630-10G
tert-Butyl-4-formylpiperidine-1-carboxylate 97%
tert-Butyl-4-formylpiperidine-1-carboxylate
Formula: C₁₁H₁₉NO₃
Melting Pt: 34…40 °C
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD02179019
CAS Number: 137076-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR