3-Bromo-2-methylthiophene

Supplier: Apollo Scientific
Warning

OR13721-1G
APOSOR13721-1GEA 21.9 EUR
APOSOR13721-1G
3-Bromo-2-methylthiophene
3-Bromo-2-methylthiophene
Formula: C₅H₅BrS
MW: 177.0622 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09033008
CAS Number: 30319-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR