Pentaethylene glycol 97%

Supplier: Apollo Scientific
Warning

Synonyms: 3,6,9,12-Tetraoxatetradecane-1,14-diol

OR13897-10G OR13897-5G OR13897-25G
APOSOR13897-10GEA 79.9 EUR
APOSOR13897-10G APOSOR13897-5G APOSOR13897-25G
Pentaethylene glycol 97%
Pentaethylene glycol
Formula: C₁₀H₂₂O₆
Boiling Pt: 338…340 °C
Density: 1,126 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002878
CAS Number: 4792-15-8
EINECS: 225-341-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR