Pyrazolo[1,5-a]pyridine-2-carboxylic acid 97+%

Supplier: Apollo Scientific
Warning

OR1408-1G
APOSOR1408-1GEA 197 EUR
APOSOR1408-1G
Pyrazolo[1,5-a]pyridine-2-carboxylic acid 97+%
Pyrazolo[1,5-a]pyridine-2-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.1500 g/mol
Melting Pt: 191…193 °C
Storage Temperature: Ambient
MDL Number: MFCD06739057
CAS Number: 63237-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR