3-Bromo-6-nitro-1H-indazole

Supplier: Apollo Scientific
Warning

OR14085-1G OR14085-5G
APOSOR14085-1GEA 21.9 EUR
APOSOR14085-1G APOSOR14085-5G
3-Bromo-6-nitro-1H-indazole
3-Bromo-6-nitro-1H-indazole
Formula: C₇H₄BrN₃O₂
Melting Pt: 233…235 °C
Storage Temperature: Ambient
MDL Number: MFCD06410881
CAS Number: 70315-68-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR