Dess-Martin periodinane

Supplier: Apollo Scientific
Warning

Synonyms: 1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one, DMP, (1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one, Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl ester

OR14263-25G OR14263-100G OR14263-250G
APOSOR14263-25GEA 29 EUR
APOSOR14263-25G APOSOR14263-100G APOSOR14263-250G
Dess-Martin periodinane
Dess-Martin periodinane
Formula: C₁₃H₁₃IO₈
MW: 424.15 g/mol
Melting Pt: 130…133 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00130127
CAS Number: 87413-09-0
UN: 1479
ADR: 5.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR