Boc-2-chloro-L-phenylalanine

Supplier: Apollo Scientific
Warning

OR14540-1G
APOSOR14540-1GEA 21.9 EUR
APOSOR14540-1G
Boc-2-chloro-L-phenylalanine
Boc-2-chloro-L-phenylalanine
Formula: C₁₄H₁₈ClNO₄
Melting Pt: 94…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00672514
CAS Number: 114873-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR