(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid 98%

Supplier: Apollo Scientific

OR14584-100G
APOSOR14584-100GEA 74.1 EUR
APOSOR14584-100G
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid
Formula: C₁₄H₁₈N₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00038128
CAS Number: 33305-77-0
EINECS: 251-450-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR