2-Bromo-6-formylpyridine 95%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Bromopyridine-2-carboxaldehyde, 6-Bromopyridine-2-carbaldehyde, 6-Bromo-2-pyridinecarboxaldehyde

OR1460-25G OR1460-100G
APOSOR1460-25GEA 55.1 EUR
APOSOR1460-25G APOSOR1460-100G
2-Bromo-6-formylpyridine 95%
2-Bromo-6-formylpyridine
Formula: C₆H₄BrNO
Melting Pt: 79…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683546
CAS Number: 34160-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR