Methyl-3-amino-6-bromobenzo[b]thiophene-2-carboxylate

Supplier: Apollo Scientific
Warning

OR14835-1G
APOSOR14835-1GEA 105 EUR
APOSOR14835-1G
Methyl-3-amino-6-bromobenzo[b]thiophene-2-carboxylate
Methyl 3-amino-6-bromobenzo[b]thiophene-2-carboxylate
Formula: C₁₀H₈BrNO₂S
Melting Pt: 126…128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09865017
CAS Number: 1017782-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR