4-Chlorobenzaldehyde oxime

Supplier: Apollo Scientific

Synonyms: 4-Chlorobenzenecarbaldehyde oxime

OR15072-1G
APOSOR15072-1GEA 21.9 EUR
APOSOR15072-1G
4-Chlorobenzaldehyde oxime
4-Chlorobenzaldehyde oxime
Formula: C₇H₆ClNO
MDL Number: MFCD00016412
CAS Number: 3848-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR