Methyl-3-hydroxy-4-methoxybenzoate

Supplier: Apollo Scientific
Warning

OR15107-1G
APOSOR15107-1GEA 21.9 EUR
APOSOR15107-1G
Methyl-3-hydroxy-4-methoxybenzoate
Methyl-3-hydroxy-4-methoxybenzoate
Formula: C₉H₁₀O₄
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD01321262
CAS Number: 6702-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR