2,5-Diphenyl-1,3-thiazol-4-ol

Supplier: Apollo Scientific
Warning

OR15188-1G
APOSOR15188-1GEA 116 EUR
APOSOR15188-1G
2,5-Diphenyl-1,3-thiazol-4-ol
2,5-Diphenyl-1,3-thiazol-4-ol
Formula: C₁₅H₁₁NOS
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00895725
CAS Number: 59484-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR