1,3-Benzoxazol-4-amine

Supplier: Apollo Scientific
Warning

Synonyms: 4-Amino-1,3-benzoxazole

OR15304-1G OR15304-5G OR15304-100MG
APOSOR15304-1GEA 85.7 EUR
APOSOR15304-1G APOSOR15304-5G APOSOR15304-100MG
1,3-Benzoxazol-4-amine
1,3-Benzoxazol-4-amine
Formula: C₇H₆N₂O
Melting Pt: 77…79 °C
Storage Temperature: Ambient
CAS Number: 163808-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR