3-Bromo-5-(hydroxymethyl)isoxazole

Supplier: Apollo Scientific
Warning

OR15590-1G OR15590-250MG
APOSOR15590-1GEA 102 EUR
APOSOR15590-1G APOSOR15590-250MG
3-Bromo-5-(hydroxymethyl)isoxazole
3-Bromo-5-(hydroxymethyl)isoxazole
Formula: C₄H₄BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD04035586
CAS Number: 25742-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR