4-Amino-2,6-dimethylpyridine

Supplier: Apollo Scientific
Warning

OR15746-1G OR15746-5G
APOSOR15746-1GEA 46.4 EUR
APOSOR15746-1G APOSOR15746-5G
4-Amino-2,6-dimethylpyridine
4-Amino-2,6-dimethylpyridine
Formula: C₇H₁₀N₂
Melting Pt: 186…187 °C
Density: 1,039 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00130078
CAS Number: 3512-80-9
EINECS: 225-515-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR