2-Chlorothiazole-4-carboxylic acid

Supplier: Apollo Scientific
Warning

OR15915-1G
APOSOR15915-1GEA 24.8 EUR
APOSOR15915-1G
2-Chlorothiazole-4-carboxylic acid
2-Chlorothiazole-4-carboxylic acid
Formula: C₄H₂ClNO₂S
Melting Pt: 220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09870033
CAS Number: 5198-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR